LT1262X1_Rev J - Large Tabletop Muffle Atmosphere Controlled Ashing_Type 30400 - Operation Manual and Parts List
The LT1262X1 Large Tabletop Muffle Atmosphere Controlled Ashing, Type 30400 - Operation Manual and Parts List covers operation and parts for various models including F30420C, F30420C-33, F30420C-33-60-80, and others. Note that Revision J includes the 3216c and 3216p controllers. This comprehensive guide is essential for understanding the operation, maintenance, and parts of these models, ensuring efficient and accurate ashing processes.
Description:
Operation manual and parts list
Note: Rev J has 3216c and 3216p controller
Environment:
Thermo Scientific Thermolyne Furnace Large Tabletop Muffle/ Atmosphere Controlled Ashing, type 30400, models:
| Models | Volts | Amps | Watts | Freq. | Phase |
| F30420C | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30420C-33 | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30420C-33-60-80 | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30420C-33-80 | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30420C-60-80 | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30420C-80 | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30430CM | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30430CM-33 | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30430CM-33-60 | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30430CM-60 | 240 | 22.9 | 5500 | 50/60 | 1 |
| F30428C | 208 | 26.4 | 5500 | 50/60 | 1 |
| F30428C-60-80 | 208 | 26.4 | 5500 | 50/60 | 1 |
| F30428C-80 | 208 | 26.4 | 5500 | 50/60 | 1 |
| F30438CM | 208 | 26.4 | 5500 | 50/60 | 1 |
| F30438CM-60 | 208 | 26.4 | 5500 | 50/60 | 1 |