Lindberg Blue M Brand Furnaces 1700C - Furnace Operations Expectations and Diagnostics
The Lindberg Blue M Brand 1700°C furnaces overview discusses their unique features, control systems, and diagnostic procedures. These furnaces use fragile molybdenum disilicide heating elements that soften when heated. They require a current-limiting device and B-Type thermocouples for temperature accuracy. Installation needs professional electrical setup. The document includes model specifics, operational expectations, and diagnostics.
Description:
Overview of the 1700°C furnaces, to better understand this equipment and some of the differences when compared to lower temperature furnaces with traditional control components. The normal operations and expectations, as well as diagnostics is covered.
Environment:
Lindberg/Blue M Brand Furnaces, 1700C, model numbers:
- BF51634C
- BF51664C
- BF51314C
- BF51524C
- STF54434C
- STF54454C
- CC59246PCOMC-1 (Control Console option for above models)
- CC59256PCOMC-1 (Control Console option for above models)
- CC59256PCM2CTC-1 (Control Console option for above models)
Attachment(s):
| File | Last Modified |
|---|---|
| Lindberg Blue M Brand Furnaces, 1700C - Furnace Operations Expectations and Diagnostics .pdf | December 17, 2021 |