Lindberg Blue M Branded Tube Furnaces HTF and STF Series - Three Zone Tube Furnace Operations and Diagnostics
The Lindberg Blue M Branded Tube Furnaces in the HTF and STF series utilize three-zone control systems for precise temperature management. Each zone, measured by individual thermocouples, is controlled using digital temperature controllers. These furnaces require a customer-supplied process tube for operation. Diagnostic procedures include checking thermocouples and controllers to ensure proper functionality. No images or attachments are included.
Description:
Instructions on operations and diagnostics of the Three Zone Tube Furnace
Environment:
Lindberg/Blue M Branded Tube Furnaces, HTF and STF series. Models numbers of Three Zone Tube Furnaces and Control Console systems:
- STF55346C-1
- STF55666C-1
- HTF55347C
- HTF55667C
- CC58434C-1
- CC58434PC-1
- CC58434PBC-1
- CC584343PC-1
- CC584343PBC-1
- STF54459C
- STF54759C
- STF54779C
- CC58475C
- CC58475PC
- CC58475PBC
- CC584753PBC
- CC58485C
- CC58485PC
- CC58485PBC
- CC584853PBC
Attachment(s):
| File | Last Modified |
|---|---|
| Lindberg Blue M Branded Tube Furnaces, HTF and STF Series - Three Zone Tube Furnace Operations and Diagnostics.pdf | December 17, 2021 |