Lindberg Blue M Furnaces - 1700C Furnace Heating Elements
The troubleshooting guide for Lindberg Blue M Furnaces - 1700C Furnace Heating Elements provides detailed instructions for addressing issues related to the heating elements. These elements are Molybdenum disilicide, ceramic-like and fragile at room temperature, but soften when heated. The resistance increases from 3-Ohms at room temperature to 6-Ohms at 1700C. A current-limiting device and a step-down transformer regulate the heater circuit. B-Type thermocouples measure temperature accurately fr
Description:
1700C Furnace Heating Elements troubleshooting guide
Environment:
Lindberg Blue M Furnaces, 1700C
Attachment(s):
| File | Last Modified |
|---|---|
| Lindberg Blue M Furnaces - 1700C Furnace Heating Elements.pdf | December 17, 2021 |