Specifications for Thermocouple, Service Part Number TC412X1A
This article describes the Thermocouple Assembly Part Number TC412X1A, used to replace the thermocouple in Barnstead Thermolyne furnaces from the F30400 Series. It is a Platinel II type thermocouple. The attachment includes probe dimensions and a list of compatible furnace models, including F30420-33-60-80, F30420C, F30420C-TS, F30420C-33, F30420C-33-80, F30420C-60-80, F30420C-80, F30428C, F30428C-60-80, F30428C-80, F30430CM, F30430CM-TS, F30430CM-33, F30430CM-33-60, F30430CM-60, and F30438CM.
Description:
Thermocouple assembly part number TC412X1A is used to replace the thermcouple in Barnstead Thermolyne furnaces from the F30400 Series.
It is a Platinel II type thermocouple.
The attachment shows probe dimensions and lists the models which feature the TC412X1A.
Environment:
F30420-33-60-80
F30420C
F30420C-TS
F30420C-33
F30420C-33-80
F30420C-60-80
F30420C-80
F30428C
F30428C-60-80
F30428C-80
F30430CM
F30430CM-TS
F30430CM-33
F30430CM-33-60
F30430CM-60
F30438CM
Attachment(s):
| File | Last Modified |
|---|---|
| Platinel II Thermcouple TC412X1A.pdf | March 18, 2025 |