056-559-00 - Rev C - Lab-Line Low Cost Ovens - Operation Manual
The "Lab-Line Low Cost Ovens - Operation Manual" covers models 3510, 3510-1, 3511, 3511-1, 3512, 3512-1, 3513, and 3513-1. It includes sections on introduction, description, specifications, installation, operation, maintenance, replacement parts, and warranty. These ovens are used for drying, sterilization, baking, and evaporation, with temperatures up to 210ºC. The manual also details power requirements and includes a warranty section.
Description:
Operation manual
Environment:
Lab-Line Low Cost Ovens, models:
| Model | Power Requirements |
| 3510 | 120 Volts, 50/60 Hz, 4.2 Amps, 500 Watts |
| 3510-1 | 240 Volts, 50/60 Hz, 2.1 Amps, 500 Watts |
| 3511 | 120 Volts, 50/60 Hz, 6.7 Amps, 800 Watts |
| 3511-1 | 240 Volts, 50/60 Hz, 3.3 Amps, 800 Watts |
| 3512 | 120 Volts, 50/60 Hz, 6.7 Amps, 800 Watts |
| 3512-1 | 240 Volts, 50/60 Hz, 3.3 Amps, 800 Watts |
| 3513 | 120 Volts, 50/60 Hz, 13.3 Amps, 1600 Watts |
| 3513-1 | 240 Volts, 50/60 Hz, 6.7 Amps, 1600 Watts |
Attachment(s):
| File | Last Modified |
|---|---|
| 056-559-00 - Rev C - Lab-Line Low Cost Ovens - Operation Manual.pdf | December 20, 2021 |