057-373-00 (7000667) - Rev 3 - Cole-Parmer Mechanical Convection Ovens - User Manual
The user manual for Cole-Parmer Mechanical Convection Ovens, document 057-373-00 (7000667) - Rev 3, covers models 52100-05 (240V) and 52100-00 (120V). It includes essential safety instructions, emphasizing the need for qualified personnel for maintenance. The ovens, suitable for various laboratory uses, feature a hydraulic thermostat, sheathed heater, constant-speed fan, and LED temperature display. The manual details unpacking, installation, maintenance, and part replacement procedures.
Description:
User manual
Environment:
Cole-Parmer Mechanical Convection Ovens, models and catalog numbers:
| Model | Catalog No. | Size | Voltage |
| 52100-05 (667) | 3515M-1C/P-Q | 1.7 cu ft | 240V |
| 52100-00 (668) | 3515MC/P-Q | 1.7 cu ft | 120V |
Attachment(s):
| File | Last Modified |
|---|---|
| 057-373-00 (7000667) - Rev 3 - Cole-Parmer Mechanical Convection Ovens - User Manual.pdf | December 21, 2021 |