107567 - Rev 0 - Thermo Scientific Hi-Temp Programmable Vacuum Oven - Operating Manual
The operating manual for the Thermo Scientific Hi-Temp Programmable Vacuum Oven provides detailed guidance on the operation, maintenance, and safety precautions for models 3625A (6250, 120V) and 3625A-1 (6251, 240V). Key features include a 304 stainless-steel work chamber, dual-electronic Safety Sentinel control system, and sheath heaters for temperature uniformity. Maintenance instructions for temperature controller, heating elements, and seals are included.
Description:
Operating manual
Environment:
Thermo Scientific Hi-Temp Programmable Vacuum Oven, models:
| Catalog numbers | Model numbers | Voltage |
| 3625A | 6250 | 120V |
| 3625A-1 | 6251 | 240V |
Attachment(s):
| File | Last Modified |
|---|---|
| 107567 - Rev 0 - Thermo Scientific Hi-Temp Programmable Vacuum Oven - Operating Manual.pdf | December 20, 2021 |