Thermo Fisher Scientific
15-3146B AU-1 - Precision Laboratory Vacuum Ovens, Models 19 and 29 - Instruction Manual
Description: 
Instruction manual
Environment:
Precision® Laboratory Vacuum Ovens, models 19 and 29. Catalog Numbers 31468, 31469, 31566, and 31568:
Catalog Number Model Volts Hz Watts Amps
31468 19 120 50/60 600 5.0
31469 19 230 50/60 600 2.6
31566 29 120 50/60 1600 13.3
31568 29 230 50/60 1600 6.9