15-3146B AU-1 - Precision Laboratory Vacuum Ovens Models 19 and 29 - Instruction Manual
The instruction manual for Precision® Laboratory Vacuum Ovens, models 19 and 29, provides detailed information on their specifications, including electrical data, and operational guidelines. Model 19 and 29 have different capacities and power requirements. The manual covers safety instructions, wiring diagrams, and features such as stainless steel chambers and vacuum seal gaskets. Model 29 includes an additional gas inlet port.
Description:
Instruction manual
Environment:
Precision® Laboratory Vacuum Ovens, models 19 and 29. Catalog Numbers 31468, 31469, 31566, and 31568:
| Catalog Number | Model | Volts | Hz | Watts | Amps |
| 31468 | 19 | 120 | 50/60 | 600 | 5.0 |
| 31469 | 19 | 230 | 50/60 | 600 | 2.6 |
| 31566 | 29 | 120 | 50/60 | 1600 | 13.3 |
| 31568 | 29 | 230 | 50/60 | 1600 | 6.9 |
Attachment(s):
| File | Last Modified |
|---|---|
| 15-3146B AU-1 - Precision Laboratory Vacuum Ovens, Models 19 and 29 - Instruction Manual.pdf | December 20, 2021 |