304068H04 - Lindberg Blue Mechanical Convection Ovens - Installation and Operation Manual
The manual for Lindberg/Blue Mechanical Convection Ovens details installation, operation, and maintenance procedures for models MO1490PA, MO1490PC, MO1490PU, MO1490SPA, MO1490SPC, and MO1490SPU. It includes safety precautions, unpacking instructions, location and wiring requirements, startup procedures, and UP150 controller operation. Maintenance covers cleaning, heating elements, blower motor, and system checks. Troubleshooting and wiring diagrams are also provided.
Description:
Installation and operation manual
Environment:
Lindberg/Blue Mechanical Convection Ovens, models:
| Model | Voltage |
| MO1490PA | 120 V, 50/60 Hz. Single Phase |
| MO1490PC | 240 V, 50/60 Hz. Single Phase |
| MO1490PU | 220 V, 50Hz Single Phase |
| MO1490SPA | 120 V, 50/60 Hz. Single Phase |
| M01490SPC | 240 V, 50/60 Hz. Single Phase |
| MO1490SPU | 220 V, 50Hz Single Phase |
Attachment(s):
| File | Last Modified |
|---|---|
| 304068H04 - Lindberg_Blue Mechanical Convection Ovens - Installation and Operation Manual.pdf | December 20, 2021 |