3177897 - Thermo Scientific Freas Micro-Controlled Ovens, Models 605, 605P, 625, and 645 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Thermo Scientific Freas Micro-Controlled Ovens, models 605, 605P, 625, and 645:
Model No. | Volts | Hz | Phase | Total Watts | Total Amps |
6050/6051 | 120 | 50/60 | 1 | 2500 | 20.8 |
6052/6053 | 230 | 50/60 | 1 | 2500 | 11 |
6054 | 230 | 50/60 | 1 | 3700 | 16 |
6056 | 230 | 50/60 | 1 | 4800 | 21 |