Thermo Fisher Scientific
3177897 - Thermo Scientific Freas Micro-Controlled Ovens, Models 605, 605P, 625, and 645 - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
Thermo Scientific Freas Micro-Controlled Ovens, models 605, 605P, 625, and 645:
Model No. Volts Hz Phase Total Watts Total Amps
6050/6051 120 50/60 1 2500 20.8
6052/6053 230 50/60 1 2500 11
6054 230 50/60 1 3700 16
6056 230 50/60 1 4800 21