3177897 - Thermo Scientific Freas Micro-Controlled Ovens_Models 605_605P_625 and 645 - Operating and Maintenance Manual
The operating and maintenance manual for the Thermo Scientific Freas Micro-Controlled Ovens, Models 605, 605P, 625, and 645, is available in different revisions, including Rev F and Rev H. These manuals provide comprehensive guidance on the operation and maintenance of the specified oven models. The manuals include detailed instructions, safety precautions, and electrical schematics. You can access these manuals through the provided links for detailed information.
Description:
Operating and maintenance manual
Environment:
Thermo Scientific Freas Micro-Controlled Ovens, models 605, 605P, 625, and 645:
| Model No. | Volts | Hz | Phase | Total Watts | Total Amps |
| 6050/6051 | 120 | 50/60 | 1 | 2500 | 20.8 |
| 6052/6053 | 230 | 50/60 | 1 | 2500 | 11 |
| 6054 | 230 | 50/60 | 1 | 3700 | 16 |
| 6056 | 230 | 50/60 | 1 | 4800 | 21 |