50041981 - Rev H - Vakuumtrockenschrank VT 6060 M-BL_VT 6130 M-BL - Betriebsanleitung
The operating manual for the vacuum drying ovens VT 6060 M-BL and VT 6130 M-BL (50041981 - Rev H) provides detailed descriptions, technical data, operating conditions, and safety warnings. The VT 6060 M-BL includes a temperature controller, inert gas connection, emergency inerting device, and two shelves. The VT 6130 M-BL is similar but has three shelves. Dimensions, weight, load capacities, and electrical specifications are provided. Safety warnings include hot surface alerts and proper socket
Description:
Betriebsanleitung
Operating manual in German
Environment:
Vakuumtrockenschrank VT 6060 M-BL / VT 6130 M-BL:
| Bestell - Nr. | Typ | Ausstattung |
| 50 014 546 | VT 6060 M - BL | Temperaturregler Digicon Temperatur-Wählbegrenzer, Inertgasan-Schluß, Not-Inertisierungseinrichtung, 2 Einlagebleche |
| 50 014 547 | VT 6130 M- BL | Temperaturregler Digicon Temperatur-Wählbegrenzer, Inertgasan-Schluß, Not-Inertisierungseinrichtung, 3 Einlagebleche |
Attachment(s):
| File | Last Modified |
|---|---|
| 50041981 - Rev H - Vakuumtrockenschrank VT 6060 M-BL VT 6130 M-BL - Betriebsanleitung.pdf | December 20, 2021 |