50041981 J - Vacuum Drying Oven VT 6060 M-BL _VT 6130 M-BL - Operating Instructions
The operating instructions for the Vacuum Drying Oven VT 6060 M-BL and VT 6130 M-BL detail their laboratory applications, including heating and vacuum processes with chamber capacities of 53 and 128 liters respectively. They operate up to 200°C and achieve vacuums of 0.01 mbar. The control panel includes various operational controls. The ovens are suited for indoor use, with specified ambient and electrical conditions.
Description:
Operating instructions
Environment:
Vacuum Drying Oven VT 6060 M-BL / VT 6130 M-BL, order numbers and models:
| Order No. | Model | Equipment |
| 50 014 546 | VT 6060 M - BL | Digicon® temperature controller, selector thermal cut-out. inen gas connection, emergency inertization facility, 2 trays |
| 50 014 547 | VT 6130 M - BL | Digicon® temperature controller, selector thermal cut-out. inert gas connection, emergency inertization facility. 3 trays |
Attachment(s):
| File | Last Modified |
|---|---|
| 50041981 J - Vacuum Drying Oven VT 6060 M-BL _ VT 6130 M-BL - Operating Instructions.pdf | December 20, 2021 |