50127995_032011 - Thermo Scientific Heratherm - Heating Coils Assembly Instructions
To assemble the heating coils for Thermo Scientific Heratherm incubators and heating/drying ovens, ensure the unit is disconnected from the power supply and follow ESD guidelines. Remove the electronics rack and rear wall, then detach the shelf system. Place the new heating coil, secure it with locking nuts, and reconnect the cables. Reassemble the unit, reconnect the power, and test the coils.
Description:
Heating coils assembly instructions
Environment:
Thermo Scientific Heratherm incubators and heating/drying ovens:
| Material Number | Description |
| 50127497 | Heating coils for Heratherm IMH-S at a voltage of 120 V |
| 50127498 | Heating coils for Heratherm IMH-S at a voltage of 230 V |
| 50127504 | Heating coils for Heratherm OMS 60, OMH 60, OMH-S 60 at a voltage of 120 V |
| 50127515 | Heating coils for Heratherm OMS 60, OMH 60, OMH-S 60 at a voltage of 230 V |
| 50127519 | Heating coils for Heratherm OMS 100, OMH 100, OMH-S 100, OMS 180, OMH 180, OMH-S 180 at a voltage betweem 208 and 240 V |
Attachment(s):
| File | Last Modified |
|---|---|
| 50127995_032011 - Thermo Scientific Heratherm - Heating Coils Assembly Instructions.pdf | December 20, 2021 |