Thermo Fisher Scientific
7177897 - Rev 0 - Freas Micro-Controlled Ovens Models 605, 605P, 625, and 645 - Installation Guide
Description: 
Installation guide
Environment:
 Freas Micro-Controlled Ovens, models 605, 605P, 625, and 645:
Model No. Volts Hz Phase Total Watts Total Amps Blower Motor Maximum Rated Temp (in °C)
6050/6051 120 50/60 1 2500 20.8 1/6 H.P. 325
6052/6053 230 50/60 1 2500 11 1/6 H.P. 325
6054 230 50/60 1 3700 16 1/6 H.P. 325
6056 230 50/60 1 4800 21 1/6 H.P. 325