7177897 - Rev 1 - Precision High Performance Ovens Models 605_605P_625_and 645 - Installation Guide
The "Precision High Performance Ovens Models 605, 605P, 625, and 645 Installation Guide" offers comprehensive instructions for installing these oven models. It covers essential steps and considerations to ensure proper setup and optimal performance. The guide is designed for service engineers and includes specific details tailored for Thermo Fisher instruments. Follow the instructions to ensure accurate installation and dependable operation of the ovens.
Description:
This installation guide contains the instructions for Precision High Performance Ovens Models 605, 605P, 625, and 645.
Environment:
Model No. Volts Hz Phase Total Watts Total Amps Blower Motor Maximum Rated Temp in C
- 6050 120 50/60 1 2500 20.8 1/6 H.P. 325
- 6052
- 6051 230 50/60 1 2500 11 1/6 H.P. 325
- 6053
- 6054 230 50/60 1 3700 16 1/6 H.P. 325
- 6056 230 50/60 1 4800 21 1/6 H.P. 325
Attachment(s):
| File | Last Modified |
|---|---|
| 7177897 - Rev 1 - Precision High Performance Ovens Models 605, 605P, 625, and 645 - Installation Guide.pdf | February 07, 2022 |