Heratherm Oven and Incubator - Main Control Board Identification and Layout
The document details the layout and electrical connections for the main control boards of Heratherm Oven and Incubator. The Standard board (Part Number 50140965) has 2 orange relays, while the High-End board (Part Number 50140962) has 3 or 4 orange relays. It includes connection points for various components such as heating elements, fans, sensors, and displays. For older units, specific instructions are provided for connecting the new PCB.
Description:
This document provides basic layout and electrical connections for General Protocol (Standard) and Advanced Protocol (High End) main control boards.
Environment:
Heratherm Oven and Incubator
Attachment(s):
| File | Last Modified |
|---|---|
| Heratherm Oven and Incubator - Main Control Board Identification and Layout.pdf | December 21, 2021 |