Heratherm_VWR_Fisher Isotemp - Power Loss Recovery in Heratherm Control Systems
This document provides how to recover from a power loss in Heratherm control systems, including VWR and Fisher Isotemp versions, follow these guidelines: For short power outages (less than 10 minutes), the unit will automatically restore full display and control the set temperature, showing error "E007". Press "esc" to clear the error. For longer outages (over 10 minutes), the unit enters standby mode, requiring a power button press to restore functionality. Advanced systems follow similar prot
Description:
Instruction sheet for power loss recovery
Environment:
Heratherm control systems including VWR and Fisher Isotemp private labeled versions
Attachment(s):
| File | Last Modified |
|---|---|
| Heratherm, VWR, Fisher Isotemp - Power Loss Recovery in Heratherm Control Systems.pdf | December 20, 2021 |