Thermo Fisher Scientific
Heratherm Oven or Incubator Overheating
Heratherm ovens or incubators may overheat due to damaged door gasket, blocked vents, excessive or dense loads, ambient heat, or incorrect setpoint. Resolution: inspect and replace faulty components, reseal door, remove obstructions, and recalibrate.
Description: 
What could cause a heratherm oven to over heat.
Environment:
OGS180
OGS100
OGS60