057-550-00 - Rev D - Thermo Scientific Lab-Line Biotronette Mark 2 Plant Growth Chambers - Operation and Repair Manual
The document is the operation and repair manual, including a parts list, for the Thermo Scientific Lab-Line Biotronette Mark 2 Plant Growth Chambers. It applies to various model numbers including 844, 845, and their variants. The manual provides detailed instructions for operating and repairing these plant growth chambers.
Description:
Operation and repair manual
Environment:
Thermo Scientific Lab-Line Biotronette Mark 2 Plant Growth Chambers, model numbers:
| 844 | 845 |
| 844-1 | 845-1 |
| 8441 | 8451 |
| 844-2 | 845-2 |
| 8442 | 8452 |
| 844P2 | 845-3 |
| 844V | 8453 |
| 844V2P2 | 845R3 |
| 845V |
- P2 With 1” Port
- V With Vapor Proof Outlet
- R3 With Recorder
Specifications:
| TEMPERATURE RANGE: |
-5°C to +55°C without lights +10°C to +55°C with lights on |
||
| TEMPERATURE CONTROL: | ±0.5°C | ||
| RELATIVE HUMIDITY RANGE: (845 MODELS ONLY) |
From ambient dew point to 70% RH, | ||
| ELECTRICAL REQUIREMENTS: | |||
| MODEL 844 | 120 VAC, Phase 1, 60 Hz, 20 Amps, 1020 Watts | ||
| MODEL 844-1 | 220 VAC, Phase 1, 60 Hz, 20 Amps, 1020 Watts | ||
| MODEL 844-2 | 220 VAC, Phase 1, 50 Hz, 20 Amps, 1020 Watts | ||
| MODEL 845 | 120 VAC, Phase 1, 60 Hz, 20 Amps; 1020 Watts | ||
| MODEL 845-1 | 220 VAC, Phase 1, 60 Hz, 20 Amps, 1020 Watts | ||
| MODEL 845-2 | 220 VAC, Phase 1, 50 Hz, 20 Amps, 1020 Watts | ||
| VOLUME: | 20 cubic feet (566 liters) | ||
Attachment(s):
| File | Last Modified |
|---|---|
| 057-550-00 - Rev D - Thermo Scientific Lab-Line Biotronette Mark 2 Plant Growth Chambers - Operation and Repair Manual.pdf | December 15, 2021 |