0600-0041-0000 - Rev E - Watlow Series SD PID Controller and PID Profiling Controller - User Manual
The document titled "Watlow Series SD PID Controller and PID Profiling Controller - User Manual" provides comprehensive instructions for the Watlow Series SD family of microprocessor-based temperature controllers. It covers various models, including static set point PID control, PID profiling ramp and soak, and FM approved limit versions, available in 1/32, 1/16, 1/8, and 1/4 DIN panel mount sizes. The manual includes details on operation, configuration, and maintenance.
Description:
User manual
Environment:
- Watlow Series SD PID Controller
- Watlow Series SD PID Profiling Controller
The Watlow Series SD family of PID, microprocessor-based temperature controllers is available in 1-32, 1-16, 1-8 and 1-4 DIN panel mount sizes. The product family includes a static set point PID control version, a PID profiling ramp and soak version and an FM approved limit version.
Attachment(s):
| File | Last Modified |
|---|---|
| 0600-0041-0000 - Rev E - Watlow Series SD PID Controller and PID Profiling Controller - User Manual.pdf | December 15, 2021 |