Thermo Fisher Scientific
50158234 - Rev B - Thermo Scientific Environmental Chamber, Model 3907 and 3948 - Operating Manual
Description: 
Operating manual
Environment:
  • Thermo Scientific Environmental Chamber, Model 3907
  • Thermo Scientific Environmental Chamber, Model 3948
Parameter Unit 3907 3948
Chamber volume L / cu ft 311 / 11 821 / 29
Chamber temperature range °C (°F) 0 - 60 (32 - 140) 0 - 60 (32 - 140)
Chamber humidity range % RH above ambient to 95 above ambient to 95