50158234 - Rev B - Thermo Scientific Environmental Chamber Model 3907 and 3948 - Operating Manual
The operating manual for the Thermo Scientific Environmental Chamber, Model 3907 and Model 3948, provides comprehensive instructions for the installation, operation, and maintenance of these environmental chambers. It ensures users can effectively manage the chambers' functionalities, adhere to safety protocols, and maintain optimal performance for various environmental testing applications.
Description:
Operating manual
Environment:
- Thermo Scientific Environmental Chamber, Model 3907
- Thermo Scientific Environmental Chamber, Model 3948
| Parameter | Unit | 3907 | 3948 |
| Chamber volume | L / cu ft | 311 / 11 | 821 / 29 |
| Chamber temperature range | °C (°F) | 0 - 60 (32 - 140) | 0 - 60 (32 - 140) |
| Chamber humidity range | % RH | above ambient to 95 | above ambient to 95 |
Attachment(s):
| File | Last Modified |
|---|---|
| 50158234 - Rev B - Thermo Scientific Environmental Chamber, Model 3907 and 3948 - Operating Manual.pdf | December 15, 2021 |