7003960 - Rev 10 - Forma Environmental Chamber, Models 3960 and 3961 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
- Forma Environmental Chamber, Model 3960, 29 cu. ft. capacity
- Forma Environmental Chamber, Model 3961, 29 cu. ft. capacity
Model 3960:
- 100-120VAC, 50/60Hz, 1 PH, 9.0 FLA
- Operating Range, including fluctuations90-125V, 50-60Hz
- 15A Breaker power switch
Model 3961:
- 200-230VAC, 50/60Hz, 1 PH, 5.0 FLA
- Operating Range, including fluctuations180-250V, 50-60Hz
- 8A Breaker power switch
Attachment(s):
File | Last Modified |
---|---|
7003960 - Rev 10 - Forma Environmental Chamber, Models 3960 and 3961 - Operating and Maintenance Manual.pdf | December 15, 2021 |