7003960 - Rev 10 - Forma Environmental Chamber Models 3960 and 3961 - Operating and Maintenance Manual
The operating and maintenance manual for the Forma Environmental Chamber, models 3960 and 3961, provides detailed instructions for the operation and maintenance of these chambers, each with a 29 cu. ft. capacity. Model 3960 operates on 100-120VAC, 50/60Hz, while model 3961 operates on 200-230VAC, 50/60Hz. The manual ensures users can effectively maintain and operate the equipment within specified voltage ranges.
Description:
Operating and maintenance manual
Environment:
- Forma Environmental Chamber, Model 3960, 29 cu. ft. capacity
- Forma Environmental Chamber, Model 3961, 29 cu. ft. capacity
Model 3960:
- 100-120VAC, 50/60Hz, 1 PH, 9.0 FLA
- Operating Range, including fluctuations90-125V, 50-60Hz
- 15A Breaker power switch
Model 3961:
- 200-230VAC, 50/60Hz, 1 PH, 5.0 FLA
- Operating Range, including fluctuations180-250V, 50-60Hz
- 8A Breaker power switch
Attachment(s):
| File | Last Modified |
|---|---|
| 7003960 - Rev 10 - Forma Environmental Chamber, Models 3960 and 3961 - Operating and Maintenance Manual.pdf | December 15, 2021 |