7023057 - Rev 1 - Portable CO2 CH-P Controller Models 3057 and 3063 - User Manual
The user manual for the Forma Scientific, Inc. Portable CO2 CH-P Controller, models 3057 and 3063, provides instructions for operation, maintenance, and troubleshooting. It includes specifications such as solid-state circuitry in a powder-coated steel casing, dimensions (12.60" W x 8.00" H x 11.40" F-B), CO2 control range (0% to 20%) with sensitivity (+/- 0.1%), and electrical requirements for each model.
Description:
User manual
Environment:
- Forma Scientific, Inc Portable CO2 CH-P Controller, model 3057
- Forma Scientific, Inc Portable CO2 CH-P Controller, model 3063
Specifications:
Construction
- Solid-state circuitry in steel casing, powder coated for a durable easily maintained surface
Dimensions
- 12.60" W x 8.00" H x 11.40" F-B (32.00cm x 20.32cm x 29.96cm)
CO2 Control
- Range: 0% TO 20%
- Control Sensitivity: +/- 0.1%
Electrical Requirements
- Model 3057: 120V, 1 Phase, 60 Hz, 1 FLA
- Model 3063: 208/230V, 1 Phase, 50 Hz, 1 FLA
Attachment(s):
| File | Last Modified |
|---|---|
| 7023057 - Rev 1 - Portable CO2 CH-P Controller, Models 3057 and 3063 - User Manual.pdf | January 20, 2022 |