Thermo Fisher Scientific
057-139-00 - Lab-Line Compact, Low Cost Incubators - Operation Manual
Description: 
Operation and repair manual and parts list
Environment:
Lab-Line Compact Low Cost Incubators, model numbers and power requirements:
Model: Power Requirements:
  • 100
  • 100IDE
  • 105
  • 120
  • 125
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 150
  • 120VAC, 50/60 Hz, 150 Watts, 1.3 Amps
  • 150JPN
  • 100VAC, 50/60 Hz, 104 Watts, 1.0 Amps
  • 101
  • 106
  • 121
  • 126
  • 240VAC, 50/60 Hz, 100 Watts, 0.4 Amps
  • 240VAC, 50/60 Hz, 100 Watts, 0.4 Amps
  • 240VAC, 50/60 Hz, 100 Watts, 0.4 Amps
  • 240VAC, 50/60 Hz, 100 Watts, 0.4 Amps
  • 152
  • 240VAC, 50/60 Hz, 150 Watts, 0.6 Amps
  • 100JPN
  • 120JPN
  • 100VAC, 50/60 Hz, 70 Watts, 0.7 Amps
  • 100VAC, 50/60 Hz, 70 Watts, 0.7 Amps
  • 100-5JPN
  • 120-5JPN
  • 100VAC, 50/60 Hz, 100 Watts, 1.0 Amps
  • 100VAC, 50/60 Hz, 100 Watts, 1.0 Amps