057-616-00 - Rev K - Lab-Line Imperial III General Purpose Incubators - Operation Manual
The document "057-616-00 - Rev K - Lab-Line Imperial III General Purpose Incubators - Operation Manual" serves as an operation and repair manual, including a parts list for various models of Lab-Line Imperial III General Purpose Incubators. The models covered include Gravity Convection and Mechanical Convection variants, each with specific features such as key locks, pen recorders, outlets, and left-hand doors.
Description:
Operation and repair manual and parts list
Environment:
Lab-Line Imperial III General Purpose Incubators, model numbers:
Gravity Convection:
- 302
- 302-1
- 302K With Key Lock
- 302R27 With 1-Pen Recorder
- 305 With Outlet
- 305-1 With Outlet
- 305L With Outlet and Left Hand Door
- 306
- 306-1
- 310 With Outlet
- 310-1 With Outlet
- 311
- 311-1
Mechanical Convection:
- 305M With Outlet
- 305M-1 With Outlet
- 306M
- 306M-1
- 310M With Outlet
- 310M-1 With Outlet
- 311M
- 311M-1
Power Requirements:
| Model No. | Power Requirements | |
| 302, 302K: | 120 V, 50/60 Hz, 2.1 amps, 250 watts | |
| 302-1: | 240 V, 50/60 Hz, 0.7 amps, 155 watts | |
| 305, 305L: | 120 V, 50/60 Hz, 3.3 amps, 400 watts | (1 ELECTRICAL OUTLET, 120 V, 500 W TOTAL LOAD) |
| 305M: | 120 V, 60 Hz, 3.3 amps, 400 watts | (1 ELECTRICAL OUTLET, 120 V, 500 W TOTAL LOAD) |
| 305-1: | 240 V, 50/60 Hz, 1.7 amps, 400 watts | (1 ELECTRICAL OUTLET, 240 V, 500 W TOTAL LOAD) |
| 305M-1: | 240 V, 50Hz, 1.7 amps, 400 watts | (1 ELECTRICAL OUTLET, 240 V, 500 W TOTAL LOAD) |
| 306: | 120 V, 50/60 Hz, 3.3 amps, 400 watts | |
| 306M: | 120 V, 60 Hz, 3.8 amps, 450 watts | |
| 306-1: | 240 V, 50/60 Hz, 1.7 amps, 400 watts | |
| 306M-1: | 240 V, 50 Hz, 1.9 amps, 450 watts | |
| 310: | 120 V, 50/60 Hz, 5.0 amps, 600 watts | (2 ELECTRICAL OUTLETS, 120 V, 500 W TOTAL LOAD) |
| 310M: | 120 V, 60 Hz, 5.0 amps, 600 watts | (2 ELECTRICAL OUTLETS, 120 V, 500 W TOTAL LOAD) |
| 310-1: | 240 V, 50/60 Hz, 2.5 amps, 600 watts | (2 ELECTRICAL OUTLETS, 240 V, 500 W TOTAL LOAD) |
| 310M-1: | 240 V, 50 Hz, 2.5 amps, 600 watts | (2 ELECTRICAL OUTLETS, 240 V, 500 W TOTAL LOAD) |
| 311: | 120 V, 50/60 Hz, 5.0 amps, 600 watts | |
| 311M: | 120 V, 50/60 Hz, 5.8 amps, 700 watts | |
| 311-1: | 240 V, 50/60 Hz, 2.5 amps, 600 watts | |
| 311M-1: | 240 V, 50/60 Hz, 2.9 amps, 700 watts |
Attachment(s):
| File | Last Modified |
|---|---|
| 057-616-00 - Rev K - Lab-Line Imperial III General Purpose Incubators - Operation Manual.pdf | December 23, 2021 |