50126461 - Heratherm Compact Microbiological Incubator IMC18 - Operating Instructions
The manual provides instructions for safe use, installation, operation, cleaning, and maintenance of the Heratherm IMC 18 compact incubator, confirming reliable microbiological culture performance.
Description:
Operating instructions
Environment:
Heratherm Compact Microbiological Incubator IMC18:
| Designation | Value |
| Volume | 18 liter |
| Dimensions (W x H x D) | 10.24 x 16.34 x 18.50 in |
| Interior dimensions | 7.87 x 12.20 x 12.20 in |
| Weight | approx. 15.87 lb |
| Minimal wall distance | 1.97 in on the sides and back |
| Number of stiffening plates | Standard 2, maximum 3 |
| Current consumption | 0.45 Amps |
| Protection/Installation class | II |
| Degree of protection | IP20 |
| Max. mechanical load | 4.41 lb per plate, 11.02 lb per device |
| Connection voltage | 100-240V AC +/-10% 50/60 Hz, Connection with the enclosed mains adapter |