50150875 - Rev D - Heratherm Refrigerated Incubators IMP 180, IMP 400 - User manual
Description:
User manual for IMP180 and IMP400 Peltier incubators
Environment:
- Heratherm Refrigerated Incubators IMP 180
- Heratherm Refrigerated Incubators IMP 400
Models | IMP 180 | IMP 400 | |
Parameter | Unit | ||
Chamber volume | L/cu ft | 178/6.3 | 381/13.5 |
Chamber temperature range | °C/°F | 5-70 (41-158) |
Attachment(s):
File | Last Modified |
---|---|
50150875_092022 Heratherm 180 400 Manual Rev D.pdf | September 18, 2023 |