50150875 - Rev D - Heratherm Refrigerated Incubators IMP 180 IMP 400 - User manual
The user manual for Heratherm Refrigerated Incubators IMP 180 and IMP 400 provides comprehensive instructions for the setup, operation, maintenance, and troubleshooting of these Peltier-incubated refrigerated incubators. The manual aims to ensure users can effectively utilize the incubators while adhering to safety standards and maintaining optimal performance.
Description:
User manual for IMP180 and IMP400 Peltier incubators
Environment:
- Heratherm Refrigerated Incubators IMP 180
- Heratherm Refrigerated Incubators IMP 400
| Models | IMP 180 | IMP 400 | |
| Parameter | Unit | ||
| Chamber volume | L/cu ft | 178/6.3 | 381/13.5 |
| Chamber temperature range | °C/°F | 5-70 (41-158) | |
Attachment(s):
| File | Last Modified |
|---|---|
| 50150875_092022 Heratherm 180 400 Manual Rev D.pdf | September 18, 2023 |