50151882 - Rev 00 - Isotemp Peltier Low Temperature Incubators - User Manual
The user manual for the Fisherbrand™ Isotemp™ Peltier Refrigerated Incubator provides detailed guidance on the installation, operation, and maintenance of the incubators. Specifically, it covers the 178L 120V model (51031568) and the 381L 120V model (51031569). The manual ensures users can effectively operate the incubators, adhere to safety protocols, and maintain optimal performance levels.
Description:
User manual for Fisherbrand™ Isotemp™ Peltier Refrigerated Incubator
Environment:
- Isotemp Peltier Low Temperature Incubator 178L 120V Model Number 51031568
- Isotemp Peltier Low Temperature Incubator 381L 120V: Model Number 51031569
| Models | Isotemp 178L | Isotemp 381L | |
| Parameter | Unit | ||
| Chamber volume | L-cu ft | 178-6.3 | 381-13.5 |
| Chamber temperature range | °C-°F | 10-60, 50-140 | |
Attachment(s):
| File | Last Modified |
|---|---|
| 50151882 - Rev 00 - Isotemp Peltier Low Temperature Incubators - User Manual.pdf | December 23, 2021 |