3177888 - Rev D - Economy Incubator - Installation Service Manual
Description:
Installation/Service Manual
Environment:
Economy incubator, models:
2EG:
- 3520
- 3521
3EG:
- 3522
- 3523
3EM:
- 3526
- 3527
5EG:
- 3524
- 3525
5EM:
- 3528
- 3529
3EM | 5EM | 2EG | 3EG | 5EG | |
Convection Technique: | Mechanical | Mechanical | Gravity | Gravity | Gravity |
Electrical Data*** (all models 50/60 Hz): | |||||
115V: | 200 W, 1.7 A | 225 W, 2.0 A | 100 W, 0.8 A | 125 W, 1.1 A | 150 W.1.3 A |
230V: | 200 W, 0.9 A | 225 W, 1.0 A | 100 W, 0.4 A | 125 W, 0.6 A | 150 W, 0.7 A |
Catalog Number, 115 V: | 3166732 | 3166734 | 3166726 | 3166728 | 3166730 |
Catalog Number, 230 V: | 3166733 | 3166735 | 3166727 | 3166729 | 3166731 |
*** All incubators equipped with line cord and plug
Attachment(s):
File | Last Modified |
---|---|
3177888 - Rev D - Economy Incubator - Installation Service Manual.pdf | January 24, 2022 |