Viewing the Heratherm Oven and Incubator Main board Firmware-Software
This article explains how to view the main board firmware-software version on Heratherm Ovens and Incubators. It details steps for both General and Advanced Protocol controllers. Accurate firmware identification is crucial for ensuring compatibility with new circulation fan motors. Firmware versions A through I expect a tachometer signal, and installing new fan designs with these firmware versions results in Error E-009. Firmware version J or higher is required to avoid this error.
Description:
Viewing the Heratherm Oven and Incubator Main board Firmware-Software
Environment:
OGS60
OGH60
OGH60
OGH60-S
OGH60-S
OGH60-S SS
OGS100
OGS100
OGH100
OGH100
OGH100-S
OGH100-S
OGH100-S SS
OGS180
OGS180
OGH180
OGH180
OGH180-S
OGH180-S
OGH180-S SS
OGS400
OGS750
OMS60
OMH60
OMH60
OMH60-S
OMH60-S
OMH60-S SS
OMS100
OMS100
OMH100
OMH100
OMH100-S
OMH100-S
OMH100-S SS
OMS180
OMS180
OMH180
OMH180
OMH180-S
OMH180-S
OMH180-S SS
OMH400
OMH400 SS
OMH750
IGS60
IGS100
IGS180
IGS400
IGS750
IMH60
IMH60
IMH60-S
IMH60-S SS
IMH100
IMH100
IMH100-S
IMH100-S SS
IMH180
IMH180
IMH180-S
IMH180-S SS
IMH400-S
IMH400-S SS
IMH750-S
IMH750SS-S
IMP180
IMP400
IMP180
IMP400
Attachment(s):
| File | Last Modified |
|---|---|
| Viewing the Heratherm Oven and Incubator Main board Firmware-Software.pdf | March 07, 2025 |