1508470 - Rev 2.1 - Thermo Scientific Multiskan Verification Plate - User Manual
Description:
User manual
Environment:
The Thermo Scientific™ Multiskan™ Verification Plate is used to verify correct measurement performance of any of the following microplate photometers:
- Thermo Scientific Multiskan FC
- Fisher Scientific™ accuSkan™ FC *
- Thermo Scientific Multiskan Ascent™**
- Thermo Scientific Multiskan EX**
* For accuSkan FC, follow the same verification instructions as for Multiskan FC.
** Discontinued
Attachment(s):
| File | Last Modified |
|---|---|
| 1508470 - Rev 2.1 - Thermo Scientific Multiskan Verification Plate - User Manual.pdf | December 28, 2021 |