9240-11-018 - Rev 0 - Compact Digital Mini Rotator, Orbital Motion - Operation Manual
Read the entire manual carefully before using the Compact Digital Mini Rotator to ensure proper operation and safety. The rotator features adjustable speeds (50-300 RPM), a timer (1 min to 99 hr 59 min), and a VFD display. Install the unit in a suitable environment, connect it to a power source, and use the provided accessories for optimal performance. Regularly clean and maintain the unit to ensure longevity.
Description:
Operation manual
Environment:
Compact Digital Mini Rotator, Orbital Motion, 100-240V AC, 50/60Hz
Attachment(s):
| File | Last Modified |
|---|---|
| 9240-11-018 - Rev 0- Compact Digital Mini Rotator, Orbital Motion - Operation Manual.pdf | December 29, 2021 |