057-621-00 - Thermo Scientific Multi-Wrist Shaker - User Manual
The Thermo Scientific Multi-Wrist Shaker User Manual provides detailed instructions for operating and maintaining the Multi-Wrist Shaker, specifically for models 3589 and 3589-1CE. It includes guidelines on setup, usage, troubleshooting, and maintenance to ensure efficient and reliable performance of the equipment.
Description:
User manual
Environment:
Thermo Scientific Multi-Wrist Shaker, model numbers:
| Model | Electrical Requirements |
| 3589 | 120 Volts, 60 Hz, 65 Watts, 0.5 Amps |
| 3589-1CE | 220/240 Volts, 50/60 Hz, 30 Watts, 0.4 Amps |
Attachment(s):
| File | Last Modified |
|---|---|
| 057-621-00 - Thermo Scientific Multi-Wrist Shaker - User Manual.pdf | December 29, 2021 |