7014535 - Rev 6 - Console Incubator Orbital Shaker - Operating Manual
The purpose of the "Console Incubator Orbital Shaker - Operating Manual" is to provide detailed operating instructions for Forma® models 4535 and 4536. These models are designed for specific electrical configurations: Model 4535 operates at a nominal 120 VAC, 60Hz, while Model 4536 operates at a nominal 230 VAC, 50 Hz. The manual ensures users understand the operating ranges and electrical requirements for safe and effective use of the equipment.
Description:
Operating manual
Environment:
Console Incubator Orbital Shaker, Forma® models:
| Model | Electrical |
| 4535 | Nominal 120 VAC, 60Hz, 1 PH, 6.5 FLA, Operating Range 90-125 VAC |
| 4536 | Nominal 230 VAC, 50 Hz, 1 PH, 3.5 FLA, Operating Range 207-253 VAC |
Attachment(s):
| File | Last Modified |
|---|---|
| 7014535 - Rev 6 - Console Incubator Orbital Shaker - Operating Manual.pdf | December 29, 2021 |