LT797X1 - Type M79700 Platform Vari-Mix Rocker - Operation Manual and Parts List
The "Type M79700 Platform Vari-Mix Rocker - Operation Manual and Parts List" provides operational instructions and a parts list for the Type M79700 Platform Vari-Mix® Rocker. It covers model numbers M79735, M79734, and M79730-26. This manual ensures users understand the operational procedures and have access to the parts information necessary for maintenance and repairs.
Description:
Operation manual and parts list
Environment:
Type M79700 Platform Vari-Mix® Rocker, model numbers:
| Model No. | Voltage | Hertz | Amps | Watts |
| M79735 | 120 | 50/60 | 0.1 | 12 |
| M79734 | 100 | 50/60 | 0.12 | 12 |
| M79730-26 | 220/240 (European only) | 50/60 | 0.05 | 12 |
Attachment(s):
| File | Last Modified |
|---|---|
| LT797X1 - Type M79700 Platform Vari-Mix Rocker - Operation Manual and Parts List.pdf | December 29, 2021 |