Thermo Fisher Scientific
9240-11-010 - Rev 0 - Digital Hotplate, HP RT2 Basic - Operation Manual
Description: 
Operation manual
Environment:
Digital Hotplate HP RT2 Basic:
Item/Model HP RT2 Basic
Dimension (W x D x H, mm) 161 x 290 x 100
Top plate dimension (Ø) 140 mm (5.5”)
Maximum heating capacity 600W
Current consumption (230V) 3A
Current consumption (120V) 5A
  • Was this article helpful?