9240-11-010 - Rev 0 - Digital Hotplate HP RT2 Basic - Operation Manual
The document "Digital Hotplate, HP RT2 Basic - Operation Manual" is an operator and user manual for Fisherbrand Isotemp RT (round top) Hot Plates. It provides detailed instructions for the operation, maintenance, and troubleshooting of the Digital Hotplate HP RT2 Basic model. The environment specifies the relevant model and catalog number: Model: 88860002 Digital Hotplate HP RT2 Basic Fisher Catalog Number: 11-676-258
Description:
Operator manual / User manual for Fisherbrand Isotemp RT (round top) Hot Plates
Environment:
88860002 Digital Hotplate HP RT2 Basic
Fisher Catalog Number 11-676-258
| Item/Model | HP RT2 Basic |
| Dimension (W x D x H, mm) | 161 x 290 x 100 |
| Top plate dimension (Ø) | 140 mm (5.5”) |
| Maximum heating capacity | 600W |
| Current consumption (230V) | 3A |
| Current consumption (120V) | 5A |
Attachment(s):
| File | Last Modified |
|---|---|
| 9240-11-010 - Rev 0 - Digital Hotplate, HP RT2 Basic - Operation Manual.pdf | December 29, 2021 |