9240-11-011 - Rev 0 - Hotplate Stirrer HPS RT2 Basic - Operation Manual
The operation manual for the Hotplate Stirrer, HPS RT2 Basic outlines the key features, specifications, and usage instructions for the device.
Description:
Operation manual
Environment:
Hotplate Stirrer, HPS RT2 Basic:
| Item/Model | HP RT2 Basic |
| Dimension (W x D x H, mm) | 161 x 290 x 100 |
| Top plate dimension (Ø) | 140 mm (5.5”) |
| Maximum heating capacity | 600W |
| Current consumption (230V) | 3A |
| Current consumption (120V) | 5A |
Attachment(s):
| File | Last Modified |
|---|---|
| 9240-11-011 - Rev 0 - Hotplate Stirrer, HPS RT2 Basic - Operation Manual.pdf | December 29, 2021 |