N11467 - Rev 1.2 - Thermo Scientific Arktik Thermal Cycler - User Manual
User Manual (Rev 1.2, 2013) for the Thermo Scientific Arktik Thermal Cycler. For professional labs, it details safety, installation, operation, and maintenance. The cycler accepts interchangeable 96, 48-dual, and 384 blocks; offers simple UI, heated lid, USB/Ethernet, and programmable protocols (gradient, touchdown, ramp control). Specs: 4–99.9 °C, 3.0 °C/s ramp, ±0.3 °C accuracy, ±0.4 °C uniformity. Includes diagnostics, troubleshooting, and ordering info.
Description:
User manual
Environment:
Thermo Scientific™ Arktik™ Thermal Cycler, catalog numbers:
- Arktik Thermal Cycler, base unit, catalog number TCA0001
- Arktik Thermal Cycler, base unit without gradient, catalog number TCA0002
Attachment(s):
| File | Last Modified |
|---|---|
| N11467 - Rev 1.2 - Thermo Scientific Arktik Thermal Cycler - User Manual.pdf | January 07, 2022 |