Thermo Fisher Scientific
Part Number for Upper Magnetic Assembly on SPD2030-230
The part number for the Upper Magnetic Assembly on the SPD2030-230 is UMA200 S.
Issue
The part number the upper magnet assembly or assembly magnet and spindle on SPD2030-230.
Environment
SPD2030-230
Assembly Magnet and Spindle (also referred to as "Upper Magnet Assembly")
Part Number: UMA200 S
Resolution
Assembly magnet and spindle is referred to as "Upper Magnet Assembly."
The part number for the Upper Magnet Assembly is UMA200 S.
  • Was this article helpful?