111-3001-00 - Rev D - Thermo Savant SpeedVac Concentrators, SC110, SC110A, SC210A - Instruction Manual
Description:
Instruction manual
Environment:
Thermo Savant centrifugal vacuum concentrators, models and electrical requirements:
- SC110 SpeedVac®
- SC110A SpeedVac® Plus
- SC210A SpeedVac® Plus
Model | Electrical Requirements |
SC110, SC110A: | 120 VAC / 60 Hz, 4 A |
230 VAC / 50 Hz, 2 A | |
SC210A: | 120 VAC / 60 Hz, 8 A |
230 VAC / 50 Hz, 4 A |
Attachment(s):
File | Last Modified |
---|---|
111-3001-00 - Rev D - Thermo Savant SpeedVac Concentrators, SC110, SC110A, SC210A - Instruction Manual.pdf | January 03, 2022 |