111-3001-00 Rev D - Thermo Savant SpeedVac Concentrators SC110_SC110A_SC210A - Instruction Manual
The Thermo Savant SpeedVac Concentrators SC110, SC110A, and SC210A instruction manual details their use in rapidly evaporating liquids to concentrate or dry samples in aqueous and organic solvents. It covers models' features, including manual and automatic bleeder valves, chamber heaters with adjustable temperatures, and various rotors for different sample types. The manual includes installation guidelines, operational procedures, and specifications, ensuring optimal use and maintenance.
Description:
Instruction manual
Environment:
Thermo Savant centrifugal vacuum concentrators, models and electrical requirements:
- SC110 SpeedVac®
- SC110A SpeedVac® Plus
- SC210A SpeedVac® Plus
| Model | Electrical Requirements |
| SC110, SC110A: | 120 VAC / 60 Hz, 4 A |
| 230 VAC / 50 Hz, 2 A | |
| SC210A: | 120 VAC / 60 Hz, 8 A |
| 230 VAC / 50 Hz, 4 A |
Attachment(s):
| File | Last Modified |
|---|---|
| 111-3001-00 - Rev D - Thermo Savant SpeedVac Concentrators, SC110, SC110A, SC210A - Instruction Manual.pdf | January 03, 2022 |