128-3000-00 - Rev D - Thermo Scientific RVT100, RVT400, RVT4104 Refrigerated Vapor Trap - Instruction Manual
Description:
Instruction manual
Environment:
Thermo Scientific® RVT100, RVT400, RVT4104 Refrigerated Vapor Trap:
Cold Traps | RVT100 | RVT400 | RVT4104 |
Operating Temperature* | -50 °C | -50 °C | -104 °C |
Capacity | 1 Liter | 4 Liter | 4 Liter |
Power Requirements | 120 VAC/60 Hz, 4 A | 120 VAC/60 Hz, 4 A | 120 VAC/60 Hz, 12 A |
240 VAC/50 Hz, 2 A | 240 VAC/50 Hz, 2 A | 240 VAC/50 Hz, 6 A | |
* Depends on ambient temperature, line voltage fluctuations, and load capacity. |
Attachment(s):
File | Last Modified |
---|---|
128-3000-00 - Rev D - Thermo Scientific RVT100, RVT400, RVT4104 Refrigerated Vapor Trap - Instruction Manual.pdf | January 03, 2022 |